ContactUsLinkComponent
Cut resistant gloves, Check & Go Red Nit 1
Cut resistant gloves, Check & Go Red Nit 1
Supplier:  HONEYWELL SAFETY
Certificates
undefined
Cut resistant gloves, Check & Go Red Nit 1
Supplier:  HONEYWELL SAFETY
Specifications
  • Catalog No:
  • 112-4677
  • DALL2332265/10
  • DALL2332265/7
  • DALL2332265/9
  • DALL2332265/8
  • Model:
  • Check & Go Red Nit 1
  • Check & Go Red Nit 1
  • Check & Go Red Nit 1
  • Check & Go Red Nit 1
  • Check & Go Red Nit 1
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Polyamide
  • Polyamide
  • Polyamide
  • Polyamide
  • Polyamide
  • Finish:
  • Palm and fingertips coated
  • Palm and fingertips coated
  • Palm and fingertips coated
  • Palm and fingertips coated
  • Palm and fingertips coated
  • Gauge:
  • 13
  • 13
  • 13
  • 13
  • 13
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (4X21A)
  • EN 388:2016 (4X21A)
  • EN 388:2016 (4X21A)
  • EN 388:2016 (4X21A)
  • EN 388:2016 (4X21A)