VWR® Buffered EDTA
Buffered EDTA, 12,5 %, 7,0, 1 L
Catalog # VWRK640901
Supplier: KLINIPATH
Jump to:
Product Details & Documents
Buffered EDTA for decalcifying fluids.
Specifications
Specifications
- Pk:1 L
- Description:Buffered EDTA, 12,5 %
- pH:7,0



