Test kits, nitrate (Testpak), CHECKIT®
Nitrate LR test kit, 0 - 1 mg/l N, 1
Catalog # LOVI147810
Supplier: Lovibond
Jump to:
Product Details & Documents
CHECKIT® Comparator is a compact and handy colorimetric unit which is suitable for both mobile and static analysis work. Supplied with a generous number of different colour scales, it provides the basis for a comprehensive, easy-to use colorimetric analysis system.
- Enables the use of large path lengths
- The mirror optics makes use of the view through the entire length of the cell
- Easy operation and exact reagent dosing
- Tablet reagents have minimum guaranteed shelf life of 5/10 years
Testpak is a simple and cost-effective means of extending the use of an existing CHECKIT® comparator instrument to a new test parameter.
: Each testpak includes the required CHECKIT® disc, tablet reagents (normally for 30 tests), two cells, stirring rod and detailed instructions for the desired method.
Specifications
Specifications
- Pk:1
- Description:Nitrate LR test kit
- Range:0 - 1 mg/l N
Specifications
Accessories
Product Information
- Catalog #PkAvailabilityPrice
- CHECKIT® plastic cells, pack of 10, with lid, 10 ml, 13,5 mm path lengthCHECKIT® plastic cells, pack of 10, with lid, 10 ml, 13,5 mm path lengthLOVI14550010Log in for availability1 * 10 ST - 43,80 €
Product Information
- Catalog #PkAvailabilityPrice
- CHECKIT® Comparator, path length 13,5 mm
Product Information
- Catalog #PkAvailabilityPrice
- CHECKIT® plastic cells, pack of 5, with lid, 10 ml, 13,5 mm path lengthCHECKIT® plastic cells, pack of 5, with lid, 10 ml, 13,5 mm path lengthLOVI1455055Log in for availability1 * 5 ST - 25,90 €
Product Information
- Catalog #PkAvailabilityPrice
- CHECKIT® plastic cells, pack of 100, with lid, 10 ml, 13,5 mm path lengthCHECKIT® plastic cells, pack of 100, with lid, 10 ml, 13,5 mm path lengthLOVI145510100Log in for availability1 * 100 ST - 363,00 €



