Test sieves, synthetic
The synthetic POM frame, nylon mesh sieves meet DIN 4197 requirements and all current amendments.
- Metal free construction
The synthetic POM frame, nylon mesh sieves meet DIN 4197 requirements and all current amendments.
- Metal free construction
Product Family Options
- ØHeightMesh size