ContactUsLinkComponent
Hood and mask, Tyvek® IsoClean®, model IC 982 B
Hood and mask, Tyvek® IsoClean®, model IC 982 B
  113-8647
 :  DuPont
undefined
Hood and mask, Tyvek® IsoClean®, model IC 982 B
  113-8647
 :  DuPont
  • Sterile:
    +
  • Garment application:
    Cleanroom
  • Colour:
    White/blue
  • Description:
    Hood and mask, 100 each per box
  • Pk:
    100
The hood is made of Tyvek® which is a fabric made of high density polyethylene using a patented flash spinning process. Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. The mask is made of pleated polyethylene.
  • Hood: Bound internal seams and head opening; ties with loops for adjustable fit; white
  • Mask: 17,5 cm wide, blue
  • Clean-processed and sterilised by gamma-irradiation to SAL of 10⁻⁶
Ideal for use in cleanrooms within the biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry as well as in other critical or controlled environments.
Suitable for use in GMP class C/D (ISO Class 6 - 9) cleanrooms, with Bioburden Control Areas.
 : Packed in a dual barrier packaging system, consisting of an inner and outer easy tear cleanroom bag, 100 per box.