ContactUsLinkComponent
Gas filters, charcoal
Gas filters, charcoal
  HEWL6810008600
 :  Agilent
Gas filters, charcoal
  HEWL6810008600
 :  Agilent
  • Description:
    Argon gas filter, small, for 700 and vista series ICP-OES
  • Pk:
    1
Gas filters are specifically designed to remove impurity gases, moisture and hydrocarbons from the ICP-OES gas supplies.
  • Deliver clean gases for accurate analyses
  • Both inlet and outlet connectors are equipped with stainless steel frits to prevent particulates from entering the gas stream
  • Easily replaced
This single combination filter removes water, trace impurity gases and organic compounds. No visual indicator. Installation date tag provided to provide a visual reminder when replacement required.