Order Entry
Belgium
ContactUsLinkComponent
VWR®, Hybridisation buffers with 50% formamide
VWR®, Hybridisation buffers with 50% formamide
Catalog # 0973-50ML
Fournisseur:  VWR Chemicals
Numéro CAS:  
undefined
VWR®, Hybridisation buffers with 50% formamide
Catalog # 0973-50ML
Fournisseur:  VWR Chemicals
Numéro CAS:  

Spécifications

  • Pk:
    50 mL
  • Description:
    Hybridisation cocktail, 50% formamide

Spécifications

À propos de cet article

Hybridisation cocktail offering blocking and hybridisation for optimised complimentary base pairing of DNA and RNA in Southern blot, Northern blot, dot blot, colony and in situ hybridisation.

Attention: Store frozen.