Orders LinkContactUsLinkComponent
Temperature stabilisation jacket
Temperature stabilisation jacket
Supplier:   SI Analytics
Temperature stabilisation jacket
Supplier:  SI Analytics
Certificates
Temperature stabilisation jacket
Temperature stabilisation jacket
4 Options Available Below
Product Details & Documents
The temperature of capillary viscometers can be stabilised in this jacket using circulation thermostats from 0 to 180 °C.
  • Support plate to facilitate the use
  • Quick-action seal to simplify the changing viscometers
  • Additional hole in the support plate so that a control thermometer can fit
Suitable for use with Ubbelohde viscometers.
Product Family Options
Product Information
  • Model
    Availability
    Price
  • SCOI285405508
    Temperature stabilisation jacket with support plate
    Temperature stabilisation jacket with support plate
    577 00
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 683,00 €
    683,00 €
    1 * 1 ST
       683,00 €
Component parts
Product Information
  • Model
    Availability
    Price
  • SCOI285405516
    Temperature stabilisation jacket
    Temperature stabilisation jacket
    577 01
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 404,00 €
    404,00 €
    1 * 1 ST
       404,00 €
  • SCOI285405532
    Silicone O-ring
    Silicone O-ring
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 21,00 €
    21,00 €
    1 * 1 ST
       21,00 €
  • SCOI285405549
    Quick-release closure
    Quick-release closure
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 91,80 €
    91,80 €
    1 * 1 ST
       91,80 €