Order Entry
Belgium
ContactUsLinkComponent
Thermostatic block heater for cuvettes, LT 20
Thermostatic block heater for cuvettes, LT 20
  HACHLTV073
 :  Hach
undefined
Thermostatic block heater for cuvettes, LT 20
  HACHLTV073
 :  Hach

 

  • Temperature Range:
    10...30 °C
  • Weight:
    4 kg
  • Description:
    Thermostatic block heater, bench-top, 80 % relative humidity, non-condensing
  • W×D×H:
    200×230×185 mm
  • Pk:
    1
  • Type:
    LT 20