ContactUsLinkComponent
TIC instrument controller, 3 head
TIC instrument controller, 3 head
  EDWAD39700000
TIC instrument controller, 3 head
  EDWAD39700000
  • Description:
    TIC instrument controller, with 3 head, RS232/RS485
  • Seq:
    01
  • Permitted ambient temperature for operation:
    0...40 °C (measured underneath TIC)
  • Power requirements:
    90 - 264 V AC, 47 - 63 Hz
  • Weight:
    45108 kg
  • Pk:
    1
TIC Instrument controller is a compact control with a large, clear graphical display, an intuitive user interface and serial communications.
  • Automatically recognises and controls active gauges, including APG100 Pirani, convection, thermocouple, strain, inverted magnetron, wide range and active ion gauges
  • Enables complete integration into PC and PLC controlled processes, all TIC variants include RS232 and RS485 interface
Windows™-based PC program provides full remote setup, control and data logging functions via the RS232 interface.
 : Accessories are sold separately. Contact your Avantor representative for more information.