Order Entry
Belgium
ContactUsLinkComponent
Solvent pumps
Solvent pumps
Catalog # MSGA16842
Supplier:  asecos
undefined
Solvent pumps
Catalog # MSGA16842
Supplier:  asecos

Specifications

  • Flow rate:
    10 l/min
  • Description:
    Solvent pump, hand operated, for containers up to 60 l
  • Immersion depth:
    600 mm
  • Pk:
    1

Specifications

About this item

Solvent pump for filling of high purity liquids, removal of organic solvents and flavourings.