Orders LinkContactUsLinkComponent
Density hydrometers, M100
Density hydrometers, M100
Supplier:   Amarell
Density hydrometers, M100
Supplier:  Amarell
Certificates
Density hydrometers, M100
Density hydrometers, M100
28 Options Available Below
Product Details & Documents
Hydrometers designed with paper scale, colour code, type designation and weighted body.
  • Round capillary
  • Accuracy of ±1 °C
  • Thermometer in the hydrometer
Suitable for government verification with (or without) mercury thermometer.
Certifications: DIN 12791
Packaging: In a transparent plastic case.
Product Family Options
Product Information
  • Model
    Range
    Division
    Length
    Availability
    Price
  • AMARH809600
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    0,600 - 0,700 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 35,50 €
    35,50 €
    1 * 1 ST
       35,50 €
  • AMARH809602
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    0,700 - 0,800 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 35,50 €
    35,50 €
    1 * 1 ST
       35,50 €
  • AMARH809604
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    0,800 - 0,900 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 35,50 €
    35,50 €
    1 * 1 ST
       35,50 €
  • AMARH809606
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    0,900 - 1,000 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 35,50 €
    35,50 €
    1 * 1 ST
       35,50 €
  • AMARH809608
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    1,000 - 1,100 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,90 €
    39,90 €
    1 * 1 ST
       39,90 €
  • AMARH809610
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    1,100 - 1,200 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,90 €
    39,90 €
    1 * 1 ST
       39,90 €
  • AMARH809612
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    1,200 - 1,300 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,90 €
    39,90 €
    1 * 1 ST
       39,90 €
  • AMARH809614
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    1,300 - 1,400 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,90 €
    42,90 €
    1 * 1 ST
       42,90 €
  • 614-0534
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    1,400 - 1,500 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,90 €
    42,90 €
    1 * 1 ST
       42,90 €
  • AMARH809618
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    1,500 - 1,600 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,90 €
    42,90 €
    1 * 1 ST
       42,90 €
  • AMARH809620
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    1,600 - 1,700 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 46,80 €
    46,80 €
    1 * 1 ST
       46,80 €
  • AMARH809622
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    1,700 - 1,800 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 46,80 €
    46,80 €
    1 * 1 ST
       46,80 €
  • AMARH809624
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    1,800 - 1,900 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 46,80 €
    46,80 €
    1 * 1 ST
       46,80 €
  • AMARH809626
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    1,900 - 2,000 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 46,80 €
    46,80 €
    1 * 1 ST
       46,80 €
Hydrometers with DKD certificate
Product Information
  • Model
    Range
    Division
    Length
    Availability
    Price
  • AMARH809604/DKD3
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    0,800 - 0,900 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • AMARH809610/DKD3
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    1,100 - 1,200 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • AMARH809612/DKD3
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    1,200 - 1,300 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • AMARH809616DKD3
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    1,400 - 1,500 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • AMARH809618DKD3
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    1,500 - 1,600 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
Hydrometers with works certificate
Product Information
  • Model
    Range
    Division
    Length
    Availability
    Price
  • AMARH809602/WC
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    0,700 - 0,800 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • AMARH809604/WC
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    0,800 - 0,900 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • AMARH809608/WC
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    1,000 - 1,100 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • AMARH809610/WC
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    1,100 - 1,200 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • AMARH809612/WC
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    1,200 - 1,300 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • AMARH809614/WC
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    1,300 - 1,400 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • AMARH809616/WC
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    1,400 - 1,500 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • AMARH809618/WC
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    1,500 - 1,600 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • AMARH809620/WC
    Density hydrometer without thermometer
    Density hydrometer without thermometer
    M100
    1,600 - 1,700 g/cm³
    0,0002 g/cm³
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST