Order Entry
Belgium
Orders LinkContactUsLinkComponent
Colour reference, RAL books for hotel

Colour reference, RAL books for hotel

Catalog #
Colour reference, RAL books for hotel
Catalog #
Supplier:  
CAS Number:  
Colour reference, RAL books for hotel
Colour reference, RAL books for hotel
CAS Number:  
Product Details & Documents

RAL book colours for hotels.

  • Coded in the RAL DESIGN system
  • Based on a scientific study, includes German/English languages
  • For architects, designers, investors, building owners, hotel managers and advertisers in the hotel industry
  • With practical and relevant design examples
  • Includes a template for colour combinations

Documents