Orders LinkContactUsLinkComponent
Illuminated surface microscopes
Illuminated surface microscopes
Supplier:   Industrial Physics
Illuminated surface microscopes
Supplier:  Industrial Physics
Certificates
Illuminated surface microscopes
4 Options Available Below
Product Details & Documents
Portable microscopes for inspectors to examine surface cleanliness, marks, pinholes, fish eyes, delaminating, cracks, and other coating defects on the spot.
  • Both scale and lens with a focusing dial
  • Surface to assess always clearly visible
  • The image you see will be inverted
Delivery information: Supplied in a black leather case.
Documents
Specifications for Product Family
Specifications
  • Catalog No:
  • SIMELD6169
  • SIMELD6170
  • SIMELD6172
  • SIMELD6174
  • Model:
  • LD6169
  • LD6170
  • LD6172
  • LD6174
  • W×D×H:
  • 45×45×45 mm
  • 175×90×55 mm
  • 175×90×55 mm
  • 175×90×55 mm
  • Weight:
  • 100 g
  • 370 g
  • 370 g
  • 370 g
Specifications
Product Family Options
Product Information
  • Magnification
    Availability
    Price
  • SIMELD6169
    Illuminated microscope with 0,10 mm graduation scale
    Illuminated microscope with 0,10 mm graduation scale
    10×
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 119,00 €
    119,00 €
    1 * 1 ST
       119,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SIMELD6169
    Model
    LD6169
    W×D×H
    45×45×45 mm
    Weight
    100 g
  • SIMELD6170
    Illuminated microscope with 0,10 mm graduation scale
    Illuminated microscope with 0,10 mm graduation scale
    20×
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 555,00 €
    555,00 €
    1 * 1 ST
       555,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SIMELD6170
    Model
    LD6170
    W×D×H
    175×90×55 mm
    Weight
    370 g
  • SIMELD6172
    Illuminated microscope with 0,02 mm graduation scale
    Illuminated microscope with 0,02 mm graduation scale
    60×
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 543,00 €
    543,00 €
    1 * 1 ST
       543,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SIMELD6172
    Model
    LD6172
    W×D×H
    175×90×55 mm
    Weight
    370 g
  • SIMELD6174
    Illuminated microscope with 0,01 mm graduation scale
    Illuminated microscope with 0,01 mm graduation scale
    100×
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 563,00 €
    563,00 €
    1 * 1 ST
       563,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SIMELD6174
    Model
    LD6174
    W×D×H
    175×90×55 mm
    Weight
    370 g