Capillary columns, DB-210
Supplier: Agilent
Jump to:
About this item
These (50%-Trifluoropropyl)-methylpolysiloxane columns are excellent for U.S. EPA Methods 8140 and 609.
- High polarity
- Bonded and cross-linked
- Solvent rinsable
- Exact replacement of HP-210
- Close equivalent to USP Phase G6
7 Options Available Below
Product Details & Documents
These (50%-Trifluoropropyl)-methylpolysiloxane columns are excellent for U.S. EPA Methods 8140 and 609.
- High polarity
- Bonded and cross-linked
- Solvent rinsable
- Exact replacement of HP-210
- Close equivalent to USP Phase G6
Product Family Options
Product Information
- Column FormatPhaseFilm thicknessØ int.LengthAvailabilityPrice
- HEWL122-0212on 7" cage
- HEWL123-0213on 7" cage
- HEWL122-0232on 7" cage
- HEWL122-0233on 7" cage
- HEWL123-0232on 7" cage
- HEWL123-0233on 7" cage
- HEWL125-0232on 7" cage