ContactUsLinkComponent
Luna® Phenyl-Hexyl Reversed Phase HPLC Columns
Luna® Phenyl-Hexyl Reversed Phase HPLC Columns
Supplier:  PHENOMENEX
Certificates
Luna® Phenyl-Hexyl Reversed Phase HPLC Columns
Supplier:  PHENOMENEX
Specifications
  • Catalog No:
  • PHEN00F-4257-E0
  • PHEN00F-4256-E0
  • PHEN00G-4257-E0
  • PHEN00F-4256-B0
  • Carbon load:
  • 17,5%
  • 17,5%
  • 17,5%
  • 17,5%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Yes
  • pH range:
  • 1,5 - 9,0
  • 1,5 - 9,0
  • 1,5 - 9,0
  • 1,5 - 9,0
  • Surface area:
  • 400 m²/g
  • 400 m²/g
  • 400 m²/g
  • 400 m²/g
  • USP code:
  • L11
  • L11
  • L11
  • L11