Order Entry
Belgium
ContactUsLinkComponent
Film applicator with reservoirs
Film applicator with reservoirs
  SIMEVF1590
undefined
Film applicator with reservoirs
  SIMEVF1590

 

  • Description:
    Film applicator, with triple reservoir, 247 g
  • Pk:
    1

 

 

Ideal for applying paint with use of a automatic film applicator or drying time recorder.

  • With three reservoirs (Ø 20 mm)
  • Can apply two pre-defined thicknesses (90 and 150 µm)

W×D×H: 25×25×90 mm