Orders LinkContactUsLinkComponent
Conductivity standards, KCL
Conductivity standards, KCL
Supplier:   Hach
Conductivity standards, KCL
Supplier:  Hach
Certificates
Conductivity standards, KCL
Conductivity standards, KCL
2 Options Available Below
Product Details & Documents
The potassium chloride standard solutions are used for calibrating conductivity cells.
Product Family Options
Product Information
  • Conductivity
    Pk
    Availability
    Price
  • HACHC20C250
    Potassium chloride standard solution, 0,1 M
    Potassium chloride standard solution, 0,1 M
    12,88 mS/cm
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 53,50 €
    53,50 €
    1 * 500 mL
       53,50 €
  • HACHC20C270
    Potassium chloride standard solution, 0,01 M
    Potassium chloride standard solution, 0,01 M
    1,413 mS/cm
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 53,50 €
    53,50 €
    1 * 500 mL
       53,50 €