Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Thickness gauges

Ultrasonic thickness gauge, Pro, 1,20 - 220 mm
  SIMELD7016
Thickness gauges
  SIMELD7016
Thickness gauges
Thickness gauges

Ultrasonic thickness gauge, Pro, 1,20 - 220 mm

 
680,00 €
   
1 * 1 ST
 

These thickness gauges are designed to measure the thickness of metallic and non-metallic materials (aluminium, titanium, plastics, ceramics, glass, plastics). It can monitor pipes and vessels for loss of thickness due to corrosion or erosion.

  • Large screen, battery indicator and a coupling indicator
  • Automatic power shut down, simple calibration procedure
  • Not suitable for cast iron due to its big crystalloid composition

'Basic' model has ten preset sound velocities.

'Pro' version allows for the storage of data (10 storage units) and performs a sound velocity measurement thanks to a known material thickness.

 : Supplied with ultrasonic gel (60 ml), probe (10 mm, 5,0 MHz), calibration block and manual. 'Pro' version has an extra probe (10 mm, 2,5 MHz).

 
  • Model:
    Pro
  • Range:
    1,20 - 220 mm
  • Description:
    Ultrasonic thickness gauge
  • Seq:
    002
  • Cat. No.:
    SIMELD7016
  • Accuracy:
    ±1,0 % H +0,1 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.