Orders LinkContactUsLinkComponent
Spanner, open end, SGE
Spanner, open end, SGE
Spanner, 3/16"×1/4" AF, Stainless steel, 3/16" and 1/4"
Catalog # 568-0686
Spanner, open end, SGE
Catalog # 568-0686
Spanner, open end, SGE
Spanner, open end, SGE
Spanner, 3/16"×1/4" AF, Stainless steel, 3/16" and 1/4"
Price
30,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
Spanner sizes and uses: 3/16" for use with mini unions, 1/4" for use with 1/16" nuts, 5/16" for use with micro control valves.
Specifications
  • Description:
    Spanner, 3/16"×1/4" AF
  • Material:
    Stainless steel
  • Use with head style:
    3/16" and 1/4"