Order Entry
Belgium
ContactUsLinkComponent
Photometer, MD 600 with reagents for waste water
Photometer, MD 600 with reagents for waste water
Catalog # LOVI214100
Supplier:  Lovibond
CAS Number:  
undefined
Photometer, MD 600 with reagents for waste water
Catalog # LOVI214100
Supplier:  Lovibond
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    Waste water setup
  • Environmentally Preferable:
  • Model:
    MD 600
  • Battery life:
    approx. 26 h
  • Wavelength accuracy:
    ±1 nm
  • Operating humidity:
    30 - 90% RH
  • Accuracy:
    2% FS (T= 20…25 °C)
  • Automatic switch-off:
    Yes
  • Compliance:
    IP 68
  • Display:
    Backlit graphic display
  • Interfaces:
    Infrared interface for data transfer
  • Operating temperature:
    5...40
  • Optics:
    LEDs, interference filters (IF) and photo sensor in transparent sample chamber
  • Power supply:
    4× AAA batteries
  • Storage capacity:
    1000 data sets
  • Wavelengths:
    430, 530, 560, 580, 610, 660 nm
  • Weight:
    ~0,45 kg
  • W×D×H:
    95×45×210 mm

Specifications

About this item

MD 600 photometer is for precise and accurate measurements with a small footprint.

  • Complete and ready to use system
  • Ideal for site laboratories where accuracy, simplicity and space are key requirements

The setup uses Lovibond ® Vario tube tests and powder packs which enable the user to easily perform highly sensitive and precise water testing. It tests COD LR(0 to 150 mg/L), COD MR(0 to 1500 mg/L), Ammonia HR(1 to 50 mg/L N), Nitrate (1 to 30 mg/L), Nitrite (0,01 to 0,3 mg/L N), Nitrogen (Total 5 to 150 mg/L N) and Phosphate (0,02 to 1 mg/L P).

Delivery: It is supplied with MD 600 photometer and its accessories, RD 125 thermoreactor, IRiM infra-red module, tube, vial stand, membrane filter set, instruction manual and guarantee sheet.