ContactUsLinkComponent
Hand brushes and brooms, PP
Hand brushes and brooms, PP
Catalog # PPSA3590300
undefined
Hand brushes and brooms, PP
Catalog # PPSA3590300
Specifications
  • Bristle colour:
    White
  • Bristle material:
    PBT
  • Description:
    Hand broom, 310×40×90 mm
  • Pk:
    1
  • Handle colour:
    White
Specifications
About this item
White hand broom with run direction for hygienically sensitive areas to clean difficult-to-access areas in cleanrooms. Material: PP (body), PBT (trimming).
  • Stock height 45 mm
  • Unslit fiber ends
  • Hard bristle material