Test kits, photometric, copper, LCK cuvette tests
Supplier: Hach
Certificates
About this item
Cuvette tests for water testing applications.
- Precise and reliable measurement values
- Maximum safety for users, thanks to the closed system and low amounts of reagents
- Convenient and error-free dosing of the reagents
- Barcode label for automatic recognition in the photometer
- Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 2 to 8 °C (keep refrigerated) for LCK529 and 15 to 25 °C for LCK229 and LCK329
Accessories information: Cuvette tests are officially approved for many legally required consent limits, with the help of Addista standard solutions and round-robin test solutions, they provide the assurance you need.
Specifications
- Environmentally Preferable:
Specifications