Order Entry
Belgium
ContactUsLinkComponent
Rhodium standard solution, for ICP-MS, Pure Plus grade
Rhodium standard solution, for ICP-MS, Pure Plus grade
Catalog # PERCN9303749
Supplier:  PerkinElmer
CAS Number:  
Rhodium standard solution, for ICP-MS, Pure Plus grade
Catalog # PERCN9303749
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    2% HCl
  • Pk:
    1
  • Pk Info:
    1× 125 ml
  • Contents:
    10 mg/l
  • Element:
    Rhodium
  • Symbol:
    Rh

Specifications

About this item

Single element standard is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.

  • Analysed by ICP-OES
  • Analysed by classical wet assay
  • 67 trace impurities analysed by ICP-MS of the final solution and reported on the certificate
  • Impurities reported at ppb level
  • Prepared and certified under ISO Guide 34 and ISO 17025 – certified by A2LA
Caution: These standards are in acid solution, for full details of hazards please read the accompanying Material Safety Data Sheets (MSDS).