Molybdenum Standard Solution,1,000 μg/ml, 2% HNO₃
Catalogus # PERCN9303745
Leverancier: PerkinElmer
CAS-nummer:
Specificaties
- Concentration:1,000 μg/ml
- Matrix:H₂O
- Pk:1
- Pk Info:125 ml
- Contents:1000 mg/l
- Element:Molybdenum
- Symbol:Mo
Specificaties
Over dit artikel
Single element standard is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.
- Analysed by ICP-OES
- Analysed by classical wet assay
- 67 trace impurities analysed by ICP-MS of the final solution and reported on the certificate
- Impurities reported at ppb level
- Prepared and certified under ISO Guide 34 and ISO 17025 – certified by A2LA
Let op: These standards are in acid solution, for full details of hazards please read the accompanying Material Safety Data Sheets (MSDS).