Order Entry
Belgium
ContactUsLinkComponent
Potassium standard solution, for ICP-MS, Pure Plus grade
Potassium standard solution, for ICP-MS, Pure Plus grade
Catalog # PERCN9303742
Supplier:  PerkinElmer
CAS Number:  
Potassium standard solution, for ICP-MS, Pure Plus grade
Catalog # PERCN9303742
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    1
  • Pk Info:
    1× 125 ml
  • Contents:
    1000 mg/l
  • Element:
    Potassium
  • Symbol:
    K

Specifications

About this item

Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.

  • Analysed by ICP-OES
  • Analysed by classical wet assay
  • 67 trace impurities analysed by ICP-MS of the final solution and reported on the certificate
  • Impurities reported at ppb level
  • Prepared and certified under ISO Guide 34 and ISO 17025 – certified by A2LA
Caution: These standards are in acid solution, for full details of hazards please read the accompanying Material Safety Data Sheets (MSDS).