Order Entry
Belgium
ContactUsLinkComponent
 
Aluminum standard solution, for AAS and ICP, Pure Plus grade
  PERCN9303726
 :  
Aluminum standard solution, for AAS and ICP, Pure Plus grade
  PERCN9303726
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    1
  • Pk Info:
    1×125 ml
  • Contents:
    1000 mg/l
  • Symbol:
    Al

 

 

Single element standard is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.

  • Analysed by ICP-OES
  • Analysed by classical wet assay
  • 32 trace impurities analysed by ICP-MS of the final solution and reported on the certificate
  • Impurities reported at ppm level
  • Prepared and certified under ISO Guide 34 and ISO 17025 – certified by A2LA
 : These standards are in acid solution, for full details of hazards please read the accompanying Material Safety Data Sheets (MSDS).