HPLC columns, ZORBAX® Eclipse XDB
Supplier: Agilent
Jump to:
About this item
Good peak shape for basic, acidic and neutral compounds.
- High performance over a wide pH range (2 to 9)
- Long lifetime with extra dens bonding and double endcapping
127 Options Available Below
Product Details & Documents
Good peak shape for basic, acidic and neutral compounds.
- High performance over a wide pH range (2 to 9)
- Long lifetime with extra dens bonding and double endcapping
Product Family Options
Product Information
- Column FormatPhasePore sizeParticle SizeØ int.LengthAvailabilityPrice
High-throughput cartridge columns (require hardware kit 820555-901)
Product Information
- Column FormatPhasePore sizeParticle SizeØ int.LengthAvailabilityPrice
- HEWL973700-906Rapid resolution cartridgeRapid resolution cartridgeGuard cartridgeEclipse XDB-C83,5 µm2,10 mm30 mmLog in for availability1 * 1 ST417,00 €
- HEWL975700-906Rapid resolution cartridgeRapid resolution cartridgeGuard cartridgeEclipse XDB-C83,5 µm2,10 mm15 mmLog in for availability1 * 1 ST
- HEWL820555-901Hardware kit
- HEWL923975-902Rapid resolution HT cartridgeRapid resolution HT cartridgeGuard cartridgeEclipse XDB-C181,8 µm4,60 mm30 mmLog in for availability1 * 1 ST593,00 €
- HEWL923700-932Rapid resolution HT cartridgeRapid resolution HT cartridgeGuard cartridgeEclipse XDB-C181,8 µm2,10 mm30 mmLog in for availability1 * 3 ST
- HEWL921700-902Rapid resolution HT cartridgeRapid resolution HT cartridgeGuard cartridgeEclipse XDB-C181,8 µm2,10 mm15 mmLog in for availability1 * 1 ST395,00 €
- HEWL921700-932Rapid resolution HT cartridgeRapid resolution HT cartridgeGuard cartridgeEclipse XDB-C181,8 µm2,10 mm15 mmLog in for availability1 * 3 ST1 010,00 €
- HEWL921975-902Rapid resolution HT cartridgeRapid resolution HT cartridgeGuard cartridgeEclipse XDB-C181,8 µm4,60 mm15 mmLog in for availability1 * 1 ST399,00 €
- HEWL921975-932Rapid resolution HT cartridgeRapid resolution HT cartridgeGuard cartridgeEclipse XDB-C181,8 µm4,60 mm15 mmLog in for availability1 * 1 ST997,00 €
- HEWL923700-902Rapid resolution HT cartridgeRapid resolution HT cartridgeGuard cartridgeEclipse XDB-C181,8 µm2,10 mm30 mmLog in for availability1 * 1 ST617,00 €
- HEWL923975-932Rapid resolution HT cartridgeRapid resolution HT cartridgeGuard cartridgeEclipse XDB-C181,8 µm4,60 mm30 mmLog in for availability1 * 3 ST1 350,00 €
- HEWL925700-902Rapid resolution HT cartridgeRapid resolution HT cartridgeGuard cartridgeEclipse XDB-C181,8 µm2,10 mm50 mmLog in for availability1 * 1 ST804,00 €
- HEWL925975-932Rapid resolution HT cartridgeRapid resolution HT cartridgeGuard cartridgeEclipse XDB-C181,8 µm4,60 mm50 mmLog in for availability1 * 1 ST1 920,00 €
- HEWL925700-932Rapid resolution HT cartridgeRapid resolution HT cartridgeGuard cartridgeEclipse XDB-C181,8 µm2,10 mm50 mmLog in for availability1 * 3 ST1 920,00 €
- HEWL933975-906Rapid resolution cartridgeRapid resolution cartridgeGuard cartridgeEclipse XDB-C83,5 µm4,60 mm30 mmLog in for availability1 * 1 ST405,00 €
- HEWL973700-902Rapid resolution cartridgeRapid resolution cartridgeGuard cartridgeEclipse XDB-C183,5 µm2,10 mm30 mmLog in for availability1 * 1 ST421,00 €
- HEWL973700-932Rapid resolution cartridgeRapid resolution cartridgeGuard cartridgeEclipse XDB-C183,5 µm2,10 mm30 mmLog in for availability1 * 3 ST1 050,00 €
- HEWL933975-902Rapid resolution cartridgeRapid resolution cartridgeGuard cartridgeEclipse XDB-C183,5 µm4,60 mm30 mmLog in for availability1 * 1 ST409,00 €
- HEWL933975-932Rapid resolution cartridgeRapid resolution cartridgeGuard cartridgeEclipse XDB-C183,5 µm4,60 mm30 mmLog in for availability1 * 3 ST1 030,00 €
- HEWL931975-906Rapid resolution cartridgeRapid resolution cartridgeGuard cartridgeEclipse XDB-C83,5 µm4,60 mm15 mmLog in for availability1 * 1 ST338,00 €
- HEWL975700-902Rapid resolution cartridgeRapid resolution cartridgeGuard cartridgeEclipse XDB-C183,5 µm2,10 mm15 mmLog in for availability1 * 1 ST328,00 €
- HEWL975700-932Rapid resolution cartridgeRapid resolution cartridgeGuard cartridgeEclipse XDB-C183,5 µm2,10 mm15 mmLog in for availability1 * 3 ST769,00 €
- HEWL931975-902Rapid resolution cartridgeRapid resolution cartridgeGuard cartridgeEclipse XDB-C183,5 µm4,60 mm15 mmLog in for availability1 * 1 ST334,00 €
- HEWL931975-932Rapid resolution cartridgeRapid resolution cartridgeGuard cartridgeEclipse XDB-C183,5 µm4,60 mm15 mmLog in for availability1 * 3 ST769,00 €
Guard columns
Product Information
- Column FormatPhasePore sizeParticle SizeØ int.LengthAvailabilityPrice
Guard cartridges
Product Information
- Column FormatPhasePore sizeParticle SizeØ int.LengthAvailabilityPrice