ContactUsLinkComponent
Inline check valves, quick-stop luer, Upchurch Scientific®
Inline check valves, quick-stop luer, Upchurch Scientific®
Catalog # 554-3155
Inline check valves, quick-stop luer, Upchurch Scientific®
Catalog # 554-3155
Specifications
  • Description:
    P-696 Quick-Stop Luer check valve assembly, 127 µl swept volume
  • Material:
    PEEK
  • Pk:
    1
Specifications
About this item
The Quick-Stop Luer check valve assembly is specifically designed to provide inline Luer connect/disconnect convenience while avoiding spills. It is made from biocompatible PEEK with gold-plated stainless steel spring.
  • Remains open when engaged
  • Designed to provide inline Luer connect/disconnect convenience without the mess and hazard of spills