Test sieves, US standard
The brass frame, stainless steel mesh sieves meet ASTM E11 requirements and all current amendments.
- Superior quality guarantees accuracy and reproducibility
- Sturdy construction ensures easy stacking
Sieve covers, made of polished brass, have a recessed wire handle.
Frequently Bought Together