Order Entry
Belgium
ContactUsLinkComponent
Calibration standard, single element, praseodymium for AAS and ICP-OES, pure grade
Calibration standard, single element, praseodymium for AAS and ICP-OES, pure grade
Catalog # PERCN9303790
Supplier:  PerkinElmer
CAS Number:  
Calibration standard, single element, praseodymium for AAS and ICP-OES, pure grade
Catalog # PERCN9303790
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    1
  • Pk Info:
    1× 125 ml
  • Element:
    Praseodymium
  • Symbol:
    Pr

Specifications

About this item

Caution: These standards are in acid solution, for full details of hazards and specification, please see website and the links to Material Safety Data Sheets (MSDS) and Technical Data sheets.