DURAN® Blanks for Distillation Flasks, Engler Type
SCOT216512805
- Ext.Ø:68 mm
- Capacity:125 ml
- Height:215 mm
- Seq:02
- Pk:100
DURAN® borosilicate glass 3.3, clear.
- With beaded rim
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Uniform wall thickness distribution makes these flasks ideal for heating applications and distillations.