Orders LinkContactUsLinkComponent
Crucibles, low form, quartz
Crucibles, low form, quartz
Crucibles, low form, quartz
Certificates
Crucibles, low form, quartz
8 Options Available Below
Product Details & Documents
Quartz, transparent, glazed.
  • Lids for low form crucible, ring handle
Ordering information: Crucible and lid have to be ordered separately.
Product Family Options
Product Information
  • Type
    Capacity
    Ø×H
    Availability
    Price
  • Low form
    6 ml
    30×19 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,50 €
    16,50 €
    1 * 1 ST
       16,50 €
  • Low form
    11 ml
    35×22 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,90 €
    18,90 €
    1 * 1 ST
       18,90 €
  • Low form
    15 ml
    41×25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,70 €
    20,70 €
    1 * 1 ST
       20,70 €
  • Low form
    25 ml
    47×28 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 21,20 €
    21,20 €
    1 * 1 ST
       21,20 €
  • Low form
    29 ml
    50×32 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 27,90 €
    27,90 €
    1 * 1 ST
       27,90 €
  • Low form
    50 ml
    60×38 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 40,10 €
    40,10 €
    1 * 1 ST
       40,10 €
  • Low form
    50 ml
    57×37 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,50 €
    39,50 €
    1 * 1 ST
       39,50 €
  • Low form
    85 ml
    70X44 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 74,40 €
    74,40 €
    1 * 1 ST
       74,40 €
Lids
Product Information
  • Catalog #
    For Ø
    Availability
    Price
  • 30 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,20 €
    28,20 €
    1 * 1 ST
       28,20 €
Product Information
  • Catalog #
    For Ø
    Availability
    Price
  • 35 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,30 €
    32,30 €
    1 * 1 ST
       32,30 €
Product Information
  • Catalog #
    For Ø
    Availability
    Price
  • 40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 41,20 €
    41,20 €
    1 * 1 ST
       41,20 €
Product Information
  • Catalog #
    For Ø
    Availability
    Price
  • 45 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,70 €
    31,70 €
    1 * 1 ST
       31,70 €
Product Information
  • Catalog #
    For Ø
    Availability
    Price
  • 50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,00 €
    43,00 €
    1 * 1 ST
       43,00 €
Product Information
  • Catalog #
    For Ø
    Availability
    Price
  • 60 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 59,10 €
    59,10 €
    1 * 1 ST
       59,10 €