Orders LinkContactUsLinkComponent
Inoculating loops, PS, calibrated
Inoculating loops, PS, calibrated
Supplier:  DELTALAB
Certificates
Inoculating loops, PS, calibrated
4 Options Available Below
Product Details & Documents
These calibrated, dual use inoculating loops offer a loop at one end to facilitate microbiological streaking, and a fine tip at the other specially designed for counting colonies.
  • High robustness and flexible
  • Colour-coded for ease of size identification
  • Eliminates the need for flaming
  • Sterilised by radiation
Product Family Options
Product Information
  • Capacity
    Colour
    Availability
    Price
  • DELT302753
    Inoculating loop, PS, sterile
    Inoculating loop, PS, sterile
    10 µl
    Blue
    Log in for availability
    Log in for availability
    1 * 1.200 ST
    1 * 1.200 ST - 186,00 €
    186,00 €
    1 * 1.200 ST
       186,00 €
  • DELT302743
    Inoculating loop, PS, sterile
    Inoculating loop, PS, sterile
    1 µl
    Green
    Log in for availability
    Log in for availability
    1 * 1.200 ST
    1 * 1.200 ST - 186,00 €
    186,00 €
    1 * 1.200 ST
       186,00 €
  • DELT302744
    Inoculating loop, PS, sterile
    Inoculating loop, PS, sterile
    1 µl
    Green
    Log in for availability
    Log in for availability
    1 * 2.800 ST
    1 * 2.800 ST - 263,00 €
    263,00 €
    1 * 2.800 ST
       263,00 €
  • DELT302754
    Inoculating loop, PS, sterile
    Inoculating loop, PS, sterile
    10 µl
    Blue
    Log in for availability
    Log in for availability
    1 * 2.800 ST
    1 * 2.800 ST - 263,00 €
    263,00 €
    1 * 2.800 ST
       263,00 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.