ContactUsLinkComponent
Acidic cleaner and neutralizer, borer 26 PLUS
  148-5266
undefined
Acidic cleaner and neutralizer, borer 26 PLUS
  148-5266
  • Description:
    borer 26 PLUS
  • Pk:
    12 kg
  • pH value:
    1,8
Liquid, highly acidic cleaning concentrate und neutraliser for automated cleaning of laboratory glassware und reusable laboratory material.
  • Rinsable without residues and guarantees consistent and error-free results
  • Neutralises carried over alkaline residues
  • Substantially reduces rinsing time and consumption of water
  • In combination with all alkaline deconex® cleaning products, can be also used as a precleaner to remove acid soluble deposits
  • Removes lime residues in laboratory washers
  • Ideal for an effective cleaning of animal cages, also suitable for use in laboratories in the dairy industry
  • EDTA-, NTA-, chlorine- and surfactant-free, no release of surfactants into waste water