ContactUsLinkComponent
Microplate handling system, StakMax®
Microplate handling system, StakMax®
Supplier:  Molecular Devices
Certificates
undefined
Microplate handling system, StakMax®
Supplier:  Molecular Devices
Specifications
  • Catalog No:
  • MLDVSTAKMAX
  • MLDV0200-6016
  • MLDV0200-6017
  • MLDV0200-6187
  • MLDV5005331
  • Model:
  • StakMax®
  • Interfaces:
  • USB 2.0
  • Plate types:
  • 96- to 384-well plates
  • Temperature operating range:
  • 10 - 40 °C
  • W×D×H:
  • 440×509×231 mm
  • Weight:
  • 11,8 kg