Order Entry
Belgium
ContactUsLinkComponent
Calibration standard, single element, tungsten for AAS and ICP-OES, pure grade
Calibration standard, single element, tungsten for AAS and ICP-OES, pure grade
Catalog # PERCN9303809
Supplier:  PerkinElmer
CAS Number:  
Calibration standard, single element, tungsten for AAS and ICP-OES, pure grade
Catalog # PERCN9303809
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pk:
    1
  • Element:
    Tungsten
  • Symbol:
    W

Specifications

About this item

Caution: These standards are in acid solution, for full details of hazards and specification, please see website and the links to Material Safety Data Sheets (MSDS) and Technical Data sheets.