ContactUsLinkComponent
Protein labelling reagent, iodoTMTzero™
Protein labelling reagent, iodoTMTzero™
Catalog # PIER90100
CAS Number:  
undefined
Protein labelling reagent, iodoTMTzero™
Catalog # PIER90100
CAS Number:  
Specifications
  • Pk:
    5 Tests
  • Description:
    iodoTMTzero™ label reagent, 5×0.2 mg
Specifications
About this item
The iodoTMTzero™ label reagent can be used as an isotopic 'light' duplex tag for MS-level protein quantitation.
  • Only reacts with sulfhydryl groups (reduced, unmodified cysteine residues), thus making it specific
  • Labeled proteins and peptides are not susceptible to reducing agents
  • Options for duplex isotopic (MS) or sixplex isobaric (MS/MS) quantitation
  • Can be used to study cysteine modifications (e.g. di-sulfide bridges, S-nitrosylation) making it versatile
This tag, along with the iodoTMTsixplex™ reagents can enable the quantitation of protein expression changes in cell-based and tissue samples that may not be amenable to metabolic isotopic labeling strategies (eg. SILAC).