ContactUsLinkComponent
Decontamination bags, activated charcoal
Decontamination bags, activated charcoal
  22007.
 :  Biotium
 :  
undefined
Decontamination bags, activated charcoal
  22007.
 :  Biotium
 :  
  • Pack type:
    bags
  • Pk:
    25
  • Description:
    Activated charcoal decontamination bags
  • Seq:
    03
Activated charcoal retains ethidium bromide, GelRed™ and GelGreen™ efficiently in a convenient bag for decontamination of used gel staining solutions. Each bag has a total capacity of at least 10 mg ethidium bromide.
  • Remove mutagenic dyes (ie, ethidium bromide) from gel staining solutions
  • Use to clean up any dyes from a solution to allow drain disposal
  • No-mess, convenient bag
  • Reusable bag capacity of at least 10 mg EtBr