Orders LinkContactUsLinkComponent
Blocking buffer, fish gelatin
Supplier:   Biotium
Blocking buffer, fish gelatin
Supplier:  Biotium
Certificates
Blocking buffer, fish gelatin
2 Options Available Below
Product Details & Documents
Fish gelatin is commonly used in blocking solutions to minimise background noise in immunodetection procedures such as Western Blotting and immunostaining. Fish gelatin can be used to block non-specific binding sites on positively charged nylon or PVDF membranes or for blocking and dilution of antibodies during immunostaining procedures. Unlike BSA or milk, fish gelatin does not contain IgG or serum proteins that could cross-react with mammalian antibodies.
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • 22010.
    Fish gelatin blocking buffer, 10% solution
    Fish gelatin blocking buffer, 10% solution
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 87,10 €
    87,10 €
    1 * 100 mL
       87,10 €
  • 22011.
    Fish gelatin powder, 2×50 µg
    Fish gelatin powder, 2×50 µg
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 68,90 €
    68,90 €
    1 * 100 g
       68,90 €