Order Entry
Belgium
Orders LinkContactUsLinkComponent
Lead(II) acetate basic ACS

Lead(II) acetate basic ACS

Irritant
Health
Environment
Lead(II) acetate basic ACS
Certificates
Lead(II) acetate basic ACS

2 Options Available Below

Product Details & Documents
Synonyms:  Horne′s compound, Lead(II) hydroxide acetate, Acetic acid lead (II) salt basic, Lead subacetate
  • CAS Number:51404-69-4
  • Formula:(CH₃COO)₂Pb·2Pb(OH)₂
  • MDL Number:MFCD00011154
  • Molecular Weight:807.72 g/mol
  • Storage Temperature:Ambient
  • UN:2291
  • ADR:6.1,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Loss on drying:
    ≤1.5 % (105°C, 2 h)
  • Titration:
    ≥33.0 % Basic lead (PbO)
  • Chloride (Cl):
    ≤0.003 %
  • Nitrate (NO3):
    ≤0.003 %
  • Calcium (Ca):
    ≤0.01 %
  • Copper (Cu):
    ≤0.002 %
  • Iron (Fe):
    ≤0.002 %
  • Potassium (K):
    ≤0.02 %
  • Sodium (Na):
    ≤0.05 %
  • Insoluble matter:
    ≤0.02 % (in dilute acetic acid)
  • Insoluble matter in water:
    ≤1.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 64,80 €
    64,80 €
    1 * 1 kg
       64,80 €

  • Plastic bottle
    5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 254,00 €
    254,00 €
    1 * 5 kg
       254,00 €