D-(+)-Glucose monohydrate 99+%, extra pure
:14431-43-7
:5996-10-1
:C₆H₁₂O₆·H₂O
:MFCD00149450
:198.17 g/mol
:83 °C
:Ambient
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Water:8 to 10 %
- HPLC:≥99.0 % (ELSD)
- Residue after ign. (sulfated):≤0.1 %
Frequently Bought Together