
Trisodium phosphate dodecahydrate, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10101-89-0
:
Formula:Na₃PO₄·12H₂O
MDL Number:MFCD00149198
:380.12 g/mol
Melting Pt:75 °C
Density:1.62 g/cm³
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to white Powder or crystals or needles
- Titration excess HCl:≥97.5 %
- Free alkali (NaOH):≤3.5 %
- Insoluble matter:≤0.05 % (in water)
Specification Test Results
Frequently Bought Together