Order Entry
Belgium
ContactUsLinkComponent
N,N-Dimethylformamide, extra dry 99.8%, AcroSeal®
Irritant
Health
Flammable
N,N-Dimethylformamide, extra dry 99.8%, AcroSeal®
Catalog # 448381000.
CAS Number:  
undefined
Irritant
Health
Flammable
N,N-Dimethylformamide, extra dry 99.8%, AcroSeal®
Catalog # 448381000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2320
  • Pk Info:
    4× 25 ml

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.005 % (Coulometric)
  • GC:
    ≥99.75 %
  • Residue after evaporation:
    ≤0.001 %
  • Filt.over 0.2 micron PFTE:
    Filtered
  • Acidity (CH3COOH):
    ≤0.002 %

Specification Test Results

About this item

 :
 :73.09 g/mol
Boiling Pt:153 °C (760 mmHg)
Melting Pt:–61 °C
Density:0.945 g/cm³
Flash Pt:58 °C
UN:2265
ADR:3,III
 :GHS07
 :GHS08
 :GHS02
Storage Temperature:Ambient
CAS Number:68-12-2
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284