Sodium chloride 99.5% for biochemistry
:7647-14-5
:
:NaCl
:MFCD00003477
:58.44 g/mol
:1461 °C (760 mmHg)
:801 °C
:2,16 g/cm³ (20 °C)
:Ambient
- Appearance:White Powder
- Titration Argentometric:≥99.4 %
- Solubility:(0.1 M in water) Clear colorless
- UV:(0.1 M in water)
- Color scale:≤15 APHA (0.1 M in water)
- pH:5.0 to 9.0
- Heavy metals (as Pb):≤0.001 %
Frequently Bought Together