Order Entry
Belgium
ContactUsLinkComponent
Ammonium sulphate 99%, pure
Ammonium sulphate 99%, pure
Certificates
undefined
Ammonium sulphate 99%, pure

Specification Test Results

  • Appearance:
    Colorless to white Crystalline powder or crystals
  • Loss on drying:
    ≤0.2 %
  • pH:
    5.0 to 6.0
  • Titration excess NaOH:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together