Order Entry
Belgium
ContactUsLinkComponent
Lithium carbonate 98+% for analysis
Irritant
Lithium carbonate 98+% for analysis
Certificates
undefined
Irritant
Lithium carbonate 98+% for analysis

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Assay:
    ≥98.0 %
  • Chloride (Cl):
    ≤0.005 %
  • Nitrate (NO3):
    ≤0.005 %
  • Sulfate (SO4):
    ≤0.3 %
  • Iron (Fe):
    ≤0.005 %
  • Sodium (Na):
    ≤0.1 %
  • Heavy metals (as Pb):
    ≤0.005 %
  • Insoluble matter:
    ≤0.05 % (in HCl)

Specification Test Results

Related Information

Frequently Bought Together