3-Hydroxy-4-methoxyphenylboronic acid pinacol ester 97%
ACRO445060010
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:White to tan Solid
- Infrared spectrum:Conforms
- Melting point:99 to 104 °C
- GC:≥96.0 %
- NMR (1H-NMR):Consistent with structure
:
:250.1 g/mol
:101…102 °C
:GHS07
:Refrigerator
:269410-23-3
:C₁₃H₁₉BO₄
:MFCD09027077