Order Entry
Belgium
ContactUsLinkComponent
Diethyl ether stabilised, Technical Grade
Irritant
Flammable
Diethyl ether stabilised, Technical Grade
Catalog # ACRO444260050
CAS Number:  
undefined
Irritant
Flammable
Diethyl ether stabilised, Technical Grade
Catalog # ACRO444260050
CAS Number:  

Specifications

  • Pack type:
    Metal drum
  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Density:
    0.713 to 0.716 kg/l
  • Stabilizer:
    5 to 10 ppm BHT

Specification Test Results

About this item

 :
 :74.12 g/mol
Boiling Pt:34.6 °C (760 mmHg)
Melting Pt:–116 °C
Density:0.714 g/cm³
Flash Pt:–45 °C
UN:1155
ADR:3,I
 :GHS07
 :GHS02
Storage Temperature:Ambient
CAS Number:60-29-7
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646