Order Entry
Belgium
ContactUsLinkComponent
 
Xylene (mixture of isomers), Technical Grade
  ACRO444240050
 :  
undefined
Xylene (mixture of isomers), Technical Grade
  ACRO444240050
 :  

 

  • Pack type:
    HDPE can, fluorinated
  • Pk:
    5 L
  • Seq:
    5190

 

 

  • Appearance:
    Clear colorless Liquid
  • Density:
    0.860 to 0.875 kg/l
  • GC:
    ≥98.5 % (Mixture of isomers)

 

 

 :
 :106.17 g/mol
 :136…140 °C (760 mmHg)
 :–34 °C
 :0.865 g/cm³
 :23 °C
 :1307
 :3,III
 :GHS02
 :GHS07
 :GHS08
 :Ambient
 :1330-20-7
 :C₆H₄(CH₃)₂
 :MFCD00077264